Assay Details
Target Gene Details
Entrez Gene ID: | 213575 |
Gene Name: | dynein cytoplasmic 2 light intermediate chain 1 |
Gene Aliases: |
4933404O11Rik, CGI-60, D2lic, LIC3, mD2LIC |
Location: |
Chr.17:84626496-84655568 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dync2li1 | NM_172256.1 | NP_758460.1 | ||
| AK008822.1 | ||||
| AK160259.1 | BAE35717.1 | |||
| BC030450.1 | AAH30450.1 | |||
| BC039070.1 | AAH39070.1 |
More Information
Set Membership: |
|

Genomic Map