Assay Details
Target Gene Details
Entrez Gene ID: | 14158 |
Gene Name: | fer (fms/fps related) protein kinase |
Gene Aliases: |
AV082135, C330004K01Rik, Fert, Fert2 |
Location: |
Chr.17:63829950-64146519 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fer | NM_001037997.3 | 9 | 1228 | NP_001033086.2 |
NM_001286415.1 | 8 | 1056 | NP_001273344.1 | |
NM_008000.2 | 2 | 145 | NP_032026.2 | |
AF286537.1 | 8 | 1094 | AAG40730.1 | |
AK082799.1 | 6 | 724 | BAC38626.1 | |
AK157828.1 | 9 | 1195 | BAE34213.1 | |
AK165973.1 | 9 | 1229 | BAE38493.1 | |
BC051249.1 | AAH51249.1 | |||
BC058100.1 | 8 | 1056 | AAH58100.1 | |
M32054.1 | 2 | 145 | AAA37617.1 | |
U76762.1 | 8 | 1161 | AAB18988.1 |
