Assay Details
Target Gene Details
Entrez Gene ID: | 75462 |
Gene Name: | RIKEN cDNA 1700001C19 gene |
Gene Aliases: |
- |
Location: |
Chr.17:47412734-47437375 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700001C19Rik | NM_029296.2 | 2 | 323 | NP_083572.1 |
| BC028263.1 | 2 | 286 | AAH28263.1 |
Target Gene Details
Entrez Gene ID: | 224833 |
Gene Name: | expressed sequence AI661453 |
Gene Aliases: |
- |
Location: |
Chr.17:47436639-47470638 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AI661453 | NM_145489.2 | NP_663464.2 | ||
| BC006605.1 | AAH06605.1 |

Genomic Map