Assay Details
Target Gene Details
Entrez Gene ID: | 75462 |
Gene Name: | RIKEN cDNA 1700001C19 gene |
Gene Aliases: |
- |
Location: |
Chr.17:47412734-47437375 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700001C19Rik | NM_029296.2 | NP_083572.1 | ||
| BC028263.1 | AAH28263.1 |

Genomic Map