Assay Details
Target Gene Details
Entrez Gene ID: | 71003 |
Gene Name: | protease, serine 41 |
Gene Aliases: |
4931440B09Rik, Tessp-1, Tessp1 |
Location: |
Chr.17:23836768-23844374 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss41 | AK049297.1 | BAC33665.1 | ||
BC150797.1 |
