Assay Details
Target Gene Details
Entrez Gene ID: | 71003 |
Gene Name: | protease, serine 41 |
Gene Aliases: |
4931440B09Rik, Tessp-1, Tessp1 |
Location: |
Chr.17:23836768-23844374 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss41 | NM_027644.1 | NP_081920.1 | ||
| AB049453.1 | BAB68561.1 | |||
| AK016509.1 | ||||
| AK049297.1 | BAC33665.1 | |||
| BC150797.1 |
More Information
Set Membership: |
|

Genomic Map