Assay Details
Target Gene Details
Entrez Gene ID: | 71003 |
Gene Name: | protease, serine 41 |
Gene Aliases: |
4931440B09Rik, Tessp-1, Tessp1 |
Location: |
Chr.17:23836768-23844374 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss41 | NM_027644.1 | 6 | 663 | NP_081920.1 |
| AB049453.1 | 6 | 663 | BAB68561.1 | |
| AK016509.1 | 6 | 641 | ||
| BC150797.1 | 6 | 679 |
More Information
Set Membership: |
|

Genomic Map