Assay Details
Target Gene Details
Entrez Gene ID: | 70544 |
Gene Name: | transmembrane protein 242 |
Gene Aliases: |
1110008A10Rik, 2310046K16Rik, 5730437N04Rik |
Location: |
Chr.17:5410864-5440260 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem242 | NM_027457.4 | NP_081733.3 | ||
| AK003559.2 | BAB22856.1 | |||
| AK009851.1 | BAC25272.1 | |||
| AK017626.1 | BAB30845.1 | |||
| AK043276.1 | ||||
| AK088025.1 | BAC40103.1 | |||
| AK155326.1 | BAE33193.1 | |||
| AK158107.1 | BAE34359.1 | |||
| BC019420.1 | AAH19420.1 | |||
| BY013779.1 |

Genomic Map