Assay Details
Target Gene Details
Entrez Gene ID: | 11682 |
Gene Name: | anaplastic lymphoma kinase |
Gene Aliases: |
CD246, Tcrz |
Location: |
Chr.17:71868988-72605410 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alk | NM_007439.2 | NP_031465.2 |
More Information
Set Membership: |
|

Genomic Map