Assay Details
Target Gene Details
Entrez Gene ID: | 225020 |
Gene Name: | fasciculation and elongation protein zeta 2 (zygin II) |
Gene Aliases: |
9030616F16, D17Ertd315e |
Location: |
Chr.17:78369212-78418152 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fez2 | NM_001285940.1 | NP_001272869.1 | ||
NM_001285946.1 | NP_001272875.1 | |||
NM_001285949.1 | NP_001272878.1 | |||
NM_199448.3 | NP_955519.1 | |||
AK033547.1 | ||||
AK150241.1 | BAE29405.1 | |||
AK157221.1 | BAE34006.1 | |||
AK159545.1 | BAE35171.1 | |||
AK165595.1 | BAE38283.1 | |||
AK218016.1 | ||||
AY382580.1 | AAR36860.1 | |||
BC064130.1 | AAH64130.1 | |||
BC096619.1 | AAH96619.1 |
