Assay Details
Target Gene Details
Entrez Gene ID: | 224727 |
Gene Name: | BCL2-associated athanogene 6 |
Gene Aliases: |
2410045D21Rik, AA408914, BAG-6, Bat3, D17H6S52E, G3, Scythe |
Location: |
Chr.17:35134600-35147322 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bag6 | NM_001252468.1 | NP_001239397.1 | ||
| NM_001252469.1 | NP_001239398.1 | |||
| NM_057171.2 | NP_476512.1 | |||
| AK010670.1 | BAB27104.1 | |||
| AK148796.1 | BAE28666.1 | |||
| AK164266.1 | ||||
| BC026647.1 | AAH26647.1 | |||
| BY595553.1 |
More Information
Set Membership: |
|

Genomic Map