Assay Details
Target Gene Details
Entrez Gene ID: | 57354 |
Gene Name: | Crm, cramped-like (Drosophila) |
Gene Aliases: |
5830477H08Rik, AI256853, AI790734, Tce4, mKIAA1426 |
Location: |
Chr.17:24961226-25015846 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cramp1l | NM_020608.3 | 18 | 3288 | NP_065633.2 |
| AK089393.1 | 18 | 3548 | ||
| AK129356.1 | 9 | 1464 | BAC98166.1 | |
| AK147411.1 | 18 | 3289 | ||
| AK147449.1 | 18 | 3289 |
More Information
Set Membership: |
|

Genomic Map