Assay Details
Target Gene Details
Entrez Gene ID: | 106369 |
Gene Name: | yippee-like 1 (Drosophila) |
Gene Aliases: |
0610009L05Rik, 1700016N17Rik, 1700019O22Rik, 4921520K19Rik, 4930511F14Rik, AV118478, Dgl1, Ppil2, mdgl-1 |
Location: |
Chr.16:17070138-17086736 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ypel1 | NM_001291047.1 | NP_001277976.1 | ||
NM_001291053.1 | NP_001277982.1 | |||
NM_023249.6 | NP_075738.1 | |||
AB098740.1 | BAD51381.1 | |||
AF190624.1 | AAG17143.1 | |||
AK005955.1 | BAB24338.1 | |||
AK006049.1 | BAB24383.2 | |||
AK006137.1 | BAC25133.1 | |||
AK006505.1 | BAB24624.1 | |||
AK006858.1 | BAB24767.1 | |||
AK011452.1 | ||||
AK014931.1 | BAB29625.1 | |||
AK015752.1 | BAB29957.1 | |||
AK018910.1 | BAB31479.1 | |||
AK019819.1 | BAB31864.1 | |||
AK076063.1 | BAC36157.1 | |||
AK133401.1 | BAE21635.1 | |||
AK139453.1 | BAE24018.1 | |||
BB610926.1 | ||||
BB649357.1 | ||||
BC049737.1 | AAH49737.1 | |||
BQ573264.1 |
