Assay Details
Target Gene Details
Entrez Gene ID: | 27409 |
Gene Name: | ATP-binding cassette, sub-family G (WHITE), member 5 |
Gene Aliases: |
AW112016, cmp, sterolin-1, trac |
Location: |
Chr.17:84658234-84682923 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcg5 | NM_031884.1 | 1 | 104 | NP_114090.1 |
| AF312713.2 | 1 | 192 | AAG53097.1 | |
| AK149569.1 | 1 | 75 | BAE28965.1 | |
| AY195872.1 | 1 | 192 | AAO45093.1 | |
| AY195873.1 | 1 | 192 | AAO45094.1 | |
| BC106766.2 | 1 | 81 | AAI06767.1 |
Target Gene Details
Entrez Gene ID: | 67470 |
Gene Name: | ATP-binding cassette, sub-family G (WHITE), member 8 |
Gene Aliases: |
1300003C16Rik, AI114946, sterolin-2 |
Location: |
Chr.17:84676302-84700333 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcg8 | NM_001286005.1 | NP_001272934.1 | ||
| AK050938.1 |
More Information
Set Membership: |
|

Genomic Map