Assay Details
Target Gene Details
Entrez Gene ID: | 27409 |
Gene Name: | ATP-binding cassette, sub-family G (WHITE), member 5 |
Gene Aliases: |
AW112016, cmp, sterolin-1, trac |
Location: |
Chr.17:84658234-84682923 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcg5 | NM_031884.1 | NP_114090.1 | ||
| AF312713.2 | AAG53097.1 | |||
| AK149569.1 | BAE28965.1 | |||
| AY195872.1 | AAO45093.1 | |||
| AY195873.1 | AAO45094.1 | |||
| BC106766.2 | AAI06767.1 |
More Information
Set Membership: |
|

Genomic Map