Assay Details
Target Gene Details
Entrez Gene ID: | 50766 |
Gene Name: | cysteine rich transmembrane BMP regulator 1 (chordin like) |
Gene Aliases: |
AU015004 |
Location: |
Chr.17:78200185-78376592 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crim1 | NM_015800.3 | 9 | 2137 | NP_056615.1 |
| AF168680.1 | 9 | 1589 | AAF34410.1 | |
| AK053197.1 | 7 | 1080 | ||
| BC100348.1 | 9 | 1922 | AAI00349.1 |

Genomic Map