Assay Details
Target Gene Details
Entrez Gene ID: | 56490 |
Gene Name: | zinc finger and BTB domain containing 20 |
Gene Aliases: |
1300017A20Rik, 7330412A13Rik, A930017C21Rik, D16Wsu73e, DPZF, HOF, ODA-8S, Oda8, Zfp288 |
Location: |
Chr.16:42875853-43621944 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb20 | NM_001285805.1 | NP_001272734.1 | ||
| NM_019778.2 | NP_062752.2 | |||
| NM_181058.1 | NP_851401.1 | |||
| AF185576.1 | AAF03152.1 | |||
| AF194030.1 | AAF06015.1 | |||
| AK005028.2 | BAB23755.1 | |||
| AK052686.1 | ||||
| BC031114.1 | AAH31114.1 | |||
| BC056446.1 | AAH56446.1 | |||
| BI659024.1 |
More Information
Set Membership: |
|

Genomic Map