Assay Details
Target Gene Details
Entrez Gene ID: | 224794 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 4 |
Gene Aliases: |
4933413N07Rik, AA986363, AI195364, E-NPP 4, Gm90, mKIAA0879 |
Location: |
Chr.17:44096307-44105924 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Enpp4 | NM_199016.2 | 3 | 1147 | NP_950181.2 |
| AK090098.1 | 3 | 1148 | BAC41090.1 | |
| AK157622.1 | 3 | 1043 | BAE34135.1 | |
| AK173048.1 | 4 | 1107 | BAD32326.1 | |
| BC027749.1 | 3 | 1101 | ||
| CB723093.1 | 2 | 596 |

Genomic Map