Assay Details
Target Gene Details
Entrez Gene ID: | 12211 |
Gene Name: | baculoviral IAP repeat-containing 6 |
Gene Aliases: |
A430032G04Rik, A430040A19Rik, AA501170, Bruce, D630005A10Rik, mKIAA1289 |
Location: |
Chr.17:74528216-74703356 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Birc6 | NM_007566.3 | 14 | 3752 | NP_031592.3 |
Y17267.1 | 14 | 3753 | CAA76720.1 |
