Assay Details
Target Gene Details
Entrez Gene ID: | 12211 |
Gene Name: | baculoviral IAP repeat-containing 6 |
Gene Aliases: |
A430032G04Rik, A430040A19Rik, AA501170, Bruce, D630005A10Rik, mKIAA1289 |
Location: |
Chr.17:74528216-74703356 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 18 - Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Birc6 | NM_007566.3 | NP_031592.3 | ||
AK134254.1 | BAE22065.1 | |||
Y17267.1 | CAA76720.1 |
