Assay Details
Target Gene Details
Entrez Gene ID: | 27762 |
Gene Name: | von Willebrand factor A domain containing 7 |
Gene Aliases: |
C6orf27, D17H6S56E-3, G7c, NG37 |
Location: |
Chr.17:35016357-35026741 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vwa7 | NM_138582.1 | 5 | 866 | NP_613048.1 |
| AF134319.1 | 5 | 866 | AAF61401.1 | |
| BC137792.1 | 5 | 807 | ||
| BC144911.1 | 5 | 807 |

Genomic Map