Assay Details
Target Gene Details
Entrez Gene ID: | 77128 |
Gene Name: | CREB3 regulatory factor |
Gene Aliases: |
A930001N09Rik, LRF |
Location: |
Chr.17:26714711-26780985 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crebrf | NM_029870.2 | NP_084146.1 | ||
AK020796.1 | BAB32214.1 | |||
AK030092.1 | BAC26779.1 | |||
AK044760.1 | BAC32070.1 | |||
AK132423.1 | ||||
AK132454.1 | ||||
BC113191.1 | ||||
BC113192.1 |
