Assay Details
Target Gene Details
Entrez Gene ID: | 77128 |
Gene Name: | CREB3 regulatory factor |
Gene Aliases: |
A930001N09Rik, LRF |
Location: |
Chr.17:26714711-26780985 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crebrf | NM_029870.2 | 5 | 1674 | NP_084146.1 |
AK030092.1 | 5 | 1675 | BAC26779.1 | |
AK132423.1 | 5 | 1647 | ||
AK132454.1 | 4 | 1420 | ||
BC113191.1 | 5 | 1568 | ||
BC113192.1 | 5 | 1568 |
