Assay Details
Target Gene Details
Entrez Gene ID: | 12211 |
Gene Name: | baculoviral IAP repeat-containing 6 |
Gene Aliases: |
A430032G04Rik, A430040A19Rik, AA501170, Bruce, D630005A10Rik, mKIAA1289 |
Location: |
Chr.17:74528216-74703356 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Birc6 | NM_007566.3 | 2 | 639 | NP_031592.3 |
| AK045446.1 | 2 | 408 | BAC32373.1 | |
| AK079995.1 | 2 | 416 | BAC37801.1 | |
| Y17267.1 | 2 | 640 | CAA76720.1 |

Genomic Map