Assay Details
Target Gene Details
Entrez Gene ID: | 106565 |
Gene Name: | delta-like 2 homolog (Drosophila) |
Gene Aliases: |
AI413481, DLK-2, Egfl9 |
Location: |
Chr.17:46297248-46303271 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dlk2 | NM_001286013.1 | NP_001272942.1 | ||
NM_001286028.1 | NP_001272957.1 | |||
BC118057.1 | ||||
BC122518.1 | ||||
BG868693.1 | ||||
FM180474.1 |
