Assay Details
Target Gene Details
Entrez Gene ID: | 106565 |
Gene Name: | delta-like 2 homolog (Drosophila) |
Gene Aliases: |
AI413481, DLK-2, Egfl9 |
Location: |
Chr.17:46297248-46303271 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlk2 | NM_001286013.1 | NP_001272942.1 | ||
| NM_001286028.1 | NP_001272957.1 | |||
| BC118057.1 | ||||
| BC122518.1 | ||||
| BG868693.1 | ||||
| BM948359.1 | ||||
| FM180474.1 |

Genomic Map