Assay Details
Target Gene Details
Entrez Gene ID: | 22661 |
Gene Name: | zinc finger protein 148 |
Gene Aliases: |
2210405J08Rik, AI480666, AW045217, BERF-1, BFCOL1, ZBP-89, Znf148 |
Location: |
Chr.16:33380775-33503903 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp148 | NM_011749.4 | NP_035879.1 | ||
| AF316548.1 | AAK63003.1 | |||
| AK036835.1 | BAC29599.1 | |||
| AK146591.1 | BAE27286.1 | |||
| BC026144.1 | AAH26144.1 | |||
| CK634070.1 | ||||
| U80078.1 | AAB38507.1 | |||
| X98096.1 | CAA66725.1 |
More Information
Set Membership: |
|

Genomic Map