Assay Details
Target Gene Details
Entrez Gene ID: | 12458 |
Gene Name: | chemokine (C-C motif) receptor 6 |
Gene Aliases: |
CC-CKR-6, CCR-6, Cmkbr6, KY411 |
Location: |
Chr.17:8236043-8257141 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccr6 | NM_001190333.1 | NP_001177262.1 | ||
| NM_001190334.1 | NP_001177263.1 | |||
| NM_001190335.1 | NP_001177264.1 | |||
| NM_001190336.1 | NP_001177265.1 | |||
| NM_001190337.1 | NP_001177266.1 | |||
| NM_001190338.1 | NP_001177267.1 | |||
| NM_009835.4 | NP_033965.1 | |||
| AB009369.1 | BAA23776.1 | |||
| AB016031.1 | BAA82443.1 | |||
| AK089712.1 | BAC40946.1 | |||
| AK154413.1 | BAE32567.1 | |||
| BC100756.1 | AAI00757.1 | |||
| BC100757.1 | AAI00758.1 | |||
| BC100758.1 | AAI00759.1 | |||
| BC105669.1 |

Genomic Map