Assay Details
Target Gene Details
Entrez Gene ID: | 110147 |
Gene Name: | euchromatic histone lysine N-methyltransferase 2 |
Gene Aliases: |
Bat8, D17Ertd710e, G9a, KMT1C, NG36 |
Location: |
Chr.17:34898464-34914051 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ehmt2 | NM_001286573.1 | 14 | 2117 | NP_001273502.1 |
| NM_001286575.1 | 16 | 2055 | NP_001273504.1 | |
| NM_145830.2 | 15 | 2219 | NP_665829.1 | |
| NM_147151.2 | 15 | 1953 | NP_671493.1 | |
| AB077209.1 | 15 | 1946 | BAC05482.1 | |
| AB077210.1 | 15 | 2219 | BAC05483.1 | |
| AK077743.1 | 3 | 275 | BAC36989.1 | |
| AK154246.1 | 14 | 2140 | BAE32462.1 | |
| BC058357.1 | 16 | 2053 | AAH58357.1 |

Genomic Map