Assay Details
Target Gene Details
Entrez Gene ID: | 17075 |
Gene Name: | epithelial cell adhesion molecule |
Gene Aliases: |
CD326, EGP, EGP-2, Egp314, Ep-CAM, EpCAM1, GA733-2, Ly74, TROP1, Tacsd1, Tacstd1, gp40 |
Location: |
Chr.17:87635979-87651127 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epcam | NM_008532.2 | NP_032558.2 | ||
AK133510.1 | ||||
AK145752.1 | BAE26627.1 | |||
AK167182.1 | BAE39317.1 | |||
BC005618.1 | AAH05618.1 | |||
BC094465.1 | AAH94465.1 | |||
M76124.1 | AAA37543.1 |
