Assay Details
Target Gene Details
Entrez Gene ID: | 12211 |
Gene Name: | baculoviral IAP repeat-containing 6 |
Gene Aliases: |
A430032G04Rik, A430040A19Rik, AA501170, Bruce, D630005A10Rik, mKIAA1289 |
Location: |
Chr.17:74528216-74703356 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Birc6 | NM_007566.3 | NP_031592.3 | ||
| AK038699.1 | 2 | 2269 | ||
| AK045446.1 | BAC32373.1 | |||
| AK079995.1 | BAC37801.1 | |||
| AK086619.1 | 1 | 1649 | ||
| Y17267.1 | CAA76720.1 |

Genomic Map