Assay Details
Target Gene Details
Entrez Gene ID: | 13163 |
Gene Name: | Fas death domain-associated protein |
Gene Aliases: |
- |
Location: |
Chr.17:33909381-33915590 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Daxx | NM_007829.4 | NP_031855.3 | ||
| DV653043.1 |

Genomic Map