Assay Details
Target Gene Details
Entrez Gene ID: | 13163 |
Gene Name: | Fas death domain-associated protein |
Gene Aliases: |
- |
Location: |
Chr.17:33909381-33915590 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Daxx | NM_001199733.1 | 2 | 447 | NP_001186662.1 |
| NM_007829.4 | 2 | 347 | NP_031855.3 | |
| AF006040.1 | 1 | 201 | AAC53284.1 | |
| AK090356.1 | 2 | 278 | ||
| AK133863.1 | 2 | 255 | BAE21895.1 | |
| AK144332.1 | 2 | 278 | ||
| AK145906.1 | 2 | 278 | BAE26741.1 | |
| AK146743.1 | 2 | 274 | BAE27403.1 | |
| AK151279.1 | 2 | 267 | ||
| AK153149.1 | 2 | 235 | BAE31759.1 | |
| AK156425.1 | 2 | 448 | BAE33708.1 | |
| AK172616.1 | 2 | 448 | ||
| BC128373.1 | 2 | 260 | ||
| DV653043.1 | 2 | 294 | ||
| KJ650207.1 | 1 | 177 |

Genomic Map