Assay Details
Target Gene Details
Entrez Gene ID: | 110147 |
Gene Name: | euchromatic histone lysine N-methyltransferase 2 |
Gene Aliases: |
Bat8, D17Ertd710e, G9a, KMT1C, NG36 |
Location: |
Chr.17:34898464-34914051 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ehmt2 | NM_001286573.1 | NP_001273502.1 | ||
| NM_001286575.1 | NP_001273504.1 | |||
| NM_145830.2 | NP_665829.1 | |||
| NM_147151.2 | NP_671493.1 | |||
| AB077209.1 | BAC05482.1 | |||
| AB077210.1 | BAC05483.1 | |||
| AK154246.1 | BAE32462.1 | |||
| AK211447.1 | ||||
| AK215390.1 | ||||
| BC058357.1 | AAH58357.1 |

Genomic Map