Assay Details
Target Gene Details
Entrez Gene ID: | 100043332 |
Gene Name: | ankyrin repeat domain 66 |
Gene Aliases: |
AU041848, D730048J04Rik, Gm4368 |
Location: |
Chr.17:43534173-43570729 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ankrd66 | NM_001254953.1 | 3 | 490 | NP_001241882.1 |
AK021352.1 | 1 | 96 |
