Assay Details
Target Gene Details
Entrez Gene ID: | 14670 |
Gene Name: | guanine nucleotide binding protein-like 1 |
Gene Aliases: |
Gna-rs1, Gnal1 |
Location: |
Chr.17:35979955-35989462 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnl1 | NM_008136.2 | NP_032162.2 | ||
AK078187.1 | ||||
BC010298.1 | AAH10298.1 | |||
BC094391.1 | AAH94391.1 | |||
BC129969.1 | ||||
BC129970.1 | ||||
BM949933.1 | ||||
X65026.1 | CAA46160.1 |
