Assay Details
Target Gene Details
Entrez Gene ID: | 56409 |
Gene Name: | nudix (nucleotide diphosphate linked moiety X)-type motif 3 |
Gene Aliases: |
1110011B09Rik, AA960325, Dipp, Dipp1 |
Location: |
Chr.17:27579382-27623452 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt3 | NM_001291046.1 | 2 | 439 | NP_001277975.1 |
| NM_019837.2 | 2 | 431 | NP_062811.1 | |
| AF264064.1 | 2 | 430 | AAF74761.1 | |
| BC016534.1 | 3 | 337 | AAH16534.1 | |
| BC046805.1 | 2 | 432 | AAH46805.1 | |
| BU919402.1 | 2 | 263 |
More Information
Set Membership: |
|

Genomic Map