Assay Details
Target Gene Details
Entrez Gene ID: | 21770 |
Gene Name: | protein phosphatase 2, regulatory subunit B', delta |
Gene Aliases: |
B'delta, TEG-271, Tex271 |
Location: |
Chr.17:46682991-46705020 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2r5d | NM_009358.3 | NP_033384.2 | ||
| AB053464.1 | BAB62015.1 | |||
| AF316616.2 | AAK01631.1 | |||
| AK154050.1 | BAE32340.1 | |||
| AK160587.1 | BAE35894.1 | |||
| AK197443.1 | ||||
| AK202461.1 | ||||
| AY338234.1 | AAQ01559.1 | |||
| BC010716.1 | AAH10716.1 |
More Information
Set Membership: |
|

Genomic Map