Assay Details
Target Gene Details
Entrez Gene ID: | 19280 |
Gene Name: | protein tyrosine phosphatase, receptor type, S |
Gene Aliases: |
AL022616, PTP, PTP-NU3, PTPNU-3, PTPsigma, Ptpt9, R-PTP-S, RPTPsigma |
Location: |
Chr.17:56412426-56476480 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprs | NM_001252453.1 | NP_001239382.1 | ||
| NM_001252455.1 | NP_001239384.1 | |||
| NM_001252456.1 | NP_001239385.1 | |||
| NM_011218.2 | NP_035348.2 | |||
| AK144123.1 | BAE25714.1 | |||
| AK159320.1 | BAE34987.1 | |||
| AK164645.1 | BAE37859.1 | |||
| AK169714.1 | BAE41325.1 | |||
| BC052462.1 | AAH52462.1 | |||
| BC083188.1 | AAH83188.1 | |||
| D28530.1 | BAA05886.1 | |||
| S59186.1 | AAB26342.2 | |||
| X82288.1 | CAA57732.1 |
More Information
Set Membership: |
|

Genomic Map