Assay Details
Target Gene Details
Entrez Gene ID: | 16004 |
Gene Name: | insulin-like growth factor 2 receptor |
Gene Aliases: |
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300 |
Location: |
Chr.17:12682406-12769706 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igf2r | NM_010515.2 | 27 | 3888 | NP_034645.2 |
| AK088699.1 | 2 | 114 | BAC40512.1 | |
| BC150817.1 | 27 | 3799 | ||
| BC171973.1 | 27 | 3799 | ||
| CA324933.1 | 4 | 384 | ||
| U04710.1 | 27 | 3844 | AAA19568.1 |
More Information
Set Membership: |
|

Genomic Map