Assay Details
Target Gene Details
Entrez Gene ID: | 16004 |
Gene Name: | insulin-like growth factor 2 receptor |
Gene Aliases: |
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300 |
Location: |
Chr.17:12682406-12769706 on Build GRCm38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igf2r | NM_010515.2 | 32 | 4662 | NP_034645.2 |
| AK088699.1 | 7 | 888 | BAC40512.1 | |
| BC150817.1 | 32 | 4573 | ||
| BC171973.1 | 32 | 4573 | ||
| CD545656.1 | 3 | 301 | ||
| CN461279.1 | 6 | 711 | ||
| U04710.1 | 32 | 4618 | AAA19568.1 |
More Information
Set Membership: |
|

Genomic Map