Assay Details
Target Gene Details
Entrez Gene ID: | 27398 |
Gene Name: | mitochondrial ribosomal protein L2 |
Gene Aliases: |
CGI-22, MRP-L14, Rpml14 |
Location: |
Chr.17:46646171-46650132 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl2 | NM_025302.3 | 4 | 523 | NP_079578.1 |
| AK007672.1 | 4 | 522 | ||
| AK009521.1 | 4 | 517 | BAB26336.1 | |
| BC027762.1 | 4 | 524 | AAH27762.1 |
More Information
Set Membership: |
|

Genomic Map