Assay Details
Target Gene Details
Entrez Gene ID: | 353344 |
Gene Name: | opsin 5 |
Gene Aliases: |
Gpr136, Neuropsin, PGR12, TMEM13 |
Location: |
Chr.17:42556779-42620470 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Opn5 | NM_181753.4 | 2 | 199 | NP_861418.2 |
| AY288426.1 | 2 | 195 | AAP72135.1 | |
| AY318865.1 | 2 | 200 | AAR08201.1 | |
| BC125611.1 | 2 | 281 | ||
| BC132010.1 | 2 | 281 |
More Information
Set Membership: |
|

Genomic Map