Assay Details
Target Gene Details
Entrez Gene ID: | 75564 |
Gene Name: | radial spoke head 9 homolog (Chlamydomonas) |
Gene Aliases: |
1700027N10Rik |
Location: |
Chr.17:46122038-46144237 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rsph9 | NM_029338.3 | 5 | 873 | NP_083614.1 |
| AK006440.1 | 5 | 876 | BAB24590.1 | |
| AK165820.1 | 5 | 744 | BAE38394.1 | |
| BC019423.1 | 5 | 847 | AAH19423.1 |
More Information
Set Membership: |
|

Genomic Map