Assay Details
Target Gene Details
Entrez Gene ID: | 16004 |
Gene Name: | insulin-like growth factor 2 receptor |
Gene Aliases: |
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300 |
Location: |
Chr.17:12682406-12769706 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 44 - Exon 44 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igf2r | NM_010515.2 | NP_034645.2 | ||
| AK053045.1 | BAC35249.1 | |||
| AK088699.1 | BAC40512.1 | |||
| BC031190.1 | AAH31190.1 | |||
| BC043457.1 | AAH43457.1 | |||
| BC055018.1 | AAH55018.1 | |||
| BC150817.1 | ||||
| BC171973.1 | ||||
| U04710.1 | AAA19568.1 |
More Information
Set Membership: |
|

Genomic Map