Assay Details
Target Gene Details
Entrez Gene ID: | 12393 |
Gene Name: | runt related transcription factor 2 |
Gene Aliases: |
AML3, Cbf, Cbfa-1, Cbfa1, LS3, Osf2, PEBP2aA, Pebp2a1, Pebpa2a |
Location: |
Chr.17:44495986-44814831 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Runx2 | NM_001146038.2 | NP_001139510.1 | ||
| NM_001271627.1 | 1 | 313 | NP_001258556.1 | |
| NM_009820.5 | NP_033950.2 | |||
| NR_073392.1 | ||||
| AF005936.1 | 1 | 267 | AAB82419.1 | |
| AF010284.1 | AAB65409.1 | |||
| AF020681.1 | AAD01649.1 | |||
| AF053951.1 | AAC78623.1 | |||
| AF053954.1 | ||||
| DQ458792.1 | ABE68713.1 |
Target Gene Details
Entrez Gene ID: | 109115 |
Gene Name: | suppressor of Ty 3 |
Gene Aliases: |
2310066G22Rik, AI315192, SPT3, SPT3L, Supt3h |
Location: |
Chr.17:44777171-45119294 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Supt3 | NM_178652.2 | NP_848767.1 | ||
| AK050321.1 | BAC34187.1 | |||
| AK075916.1 | BAC36052.1 | |||
| AK087935.1 | BAC40048.1 | |||
| AK089837.1 |
More Information
Set Membership: |
|

Genomic Map