Assay Details
Target Gene Details
Entrez Gene ID: | 106581 |
Gene Name: | integrin alpha FG-GAP repeat containing 3 |
Gene Aliases: |
- |
Location: |
Chr.17:26212692-26244242 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itfg3 | NM_001206335.1 | 12 | 1793 | NP_001193264.1 |
NM_207217.4 | 11 | 1681 | NP_997100.1 | |
AK029427.1 | 11 | 1581 | BAC26447.1 | |
AK170646.1 | 12 | 1712 | BAE41933.1 | |
BC046457.1 | 12 | 1640 | AAH46457.1 |
