Assay Details
Target Gene Details
Entrez Gene ID: | 68565 |
Gene Name: | mitochondrial ribosomal protein S18A |
Gene Aliases: |
1110004O20Rik, C79712 |
Location: |
Chr.17:46111004-46128908 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps18a | NM_026768.3 | NP_081044.1 | ||
| AB049953.1 | BAB41006.1 | |||
| AK003444.1 | BAB22794.1 | |||
| AK028226.1 | BAC25825.1 | |||
| AK172171.1 | BAE42864.1 | |||
| BC025456.1 | AAH25456.1 | |||
| BC058959.1 | AAH58959.1 |
Target Gene Details
Entrez Gene ID: | 75564 |
Gene Name: | radial spoke head 9 homolog (Chlamydomonas) |
Gene Aliases: |
1700027N10Rik |
Location: |
Chr.17:46122038-46144237 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rsph9 |
More Information
Set Membership: |
|

Genomic Map