Assay Details
Target Gene Details
Entrez Gene ID: | 18771 |
Gene Name: | Pbx/knotted 1 homeobox |
Gene Aliases: |
D17Wsu76e, PREP1 |
Location: |
Chr.17:31564721-31607693 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pknox1 | NM_016670.3 | NP_057879.2 | ||
| NR_027493.1 | ||||
| AF061270.1 | AAC15990.1 | |||
| AK131788.1 | BAE20805.1 | |||
| AK134000.1 | BAE21975.1 | |||
| BC052701.1 | AAH52701.1 |
More Information
Set Membership: |
|

Genomic Map