Assay Details
Target Gene Details
Entrez Gene ID: | 224902 |
Gene Name: | scaffold attachment factor B2 |
Gene Aliases: |
AA389433, AI255170, mKIAA0138 |
Location: |
Chr.17:56560971-56584672 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Safb2 | NM_001029979.2 | NP_001025150.2 | ||
| AK031971.1 | BAC27631.1 | |||
| BC028827.1 | AAH28827.1 | |||
| BC050855.1 | AAH50855.1 | |||
| BC107016.2 |

Genomic Map